Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MLVTHDYTAV KEDEINVYQG EVVQILASNQ QNMFLVFRAA TDQCPAAEGW IPGFVLGHTS AVIVENPDGT LKKSTSWHTA LRLRKKSEKK DKDGKREGKL ENGYRKSREG LSNKVSVKLL NPN
Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.
TRIO promotes the exchange of GDP by GTP. Together with leukocyte antigen-related (LAR) protein, it could play a role in coordinating cell-matrix and cytoskeletal rearrangements necessary for cell migration and cell growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PTPRF-interacting protein; triple functional domain (PTPRF interacting); Triple functional domain protein
Gene Aliases: ARHGEF23; tgat; TRIO
UniProt ID: (Human) O75962
Entrez Gene ID: (Human) 7204
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support