Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MSGPTWLPPK QPEPARAPQG RAIPRGTPGP PPAHGAALQP HPRVNFCPLP SEQCYQAPGG PEDRGPAWVG SHGVLQHTQG LPADRGGLRP GSLDAEIDLL SSTLAELNGG RGHASRRPDR QAYEPPPPPA YRTGSLKPNP ASPLPASPYG GPTPASYTTA STPAGPAFPV QVKVAQPVRG CGPPRRGASQ ASGPLPGPHF PLPGRGEVWG PGYRSQREPG PGAKEEAAGV SGPAGRGRGG EHGPQVPLSQ PPEDELDRLT KKLVHDMNHP PSGEYFGQCG GCGEDVVGDG AGVVALDRVF HVGCFVCSTC RAQLRGQHFY AVERRAYCEG CYVATLEKCA TCSQPILDRI LRAMGKAYHP GCFTCVVCHR GLDGIPFTVD ATSQIHCIED FHRKFAPRCS VCGGAIMPEP GQEETVRIVA LDRSFHIGCY KCEECGLLLS SEGECQGCYP LDGHILCKAC SAWRIQELSA TVTTDC
This gene is a member of the zyxin family and encodes a protein with three LIM zinc-binding domains. This protein localizes to focal adhesion sites and along actin stress fibers. Recruitment of this protein to the plasma membrane occurs in a lysophosphatidic acid (LPA)-dependent manner and it regulates LPA-induced cell migration. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC10556; MGC10558; MGC29959; MGC3837; MGC4423; OIP-1; OPA-interacting protein 1; tcag7.1300; thyroid hormone receptor interacting protein 6; Thyroid receptor-interacting protein 6; TR-interacting protein 6; ZRP-1; zyxin related protein 1; Zyxin-related protein 1
Gene Aliases: OIP-1; OIP1; TRIP-6; TRIP6; TRIP6i2; ZRP-1
UniProt ID: (Human) Q15654
Entrez Gene ID: (Human) 7205
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support