Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MA5-17191 targets TRIP6 in FACS, pep-ELISA, and WB applications and shows reactivity with Human samples.
The MA5-17191 immunogen is purified recombinant fragment of human TRIP6 (amino acids: 107-291) expressed in E. Coli.
MA5-17191 detects TRIP6 which has a predicted molecular weight of approximately 50.3kDa.
This gene is a member of the zyxin family and encodes a protein with three LIM zinc-binding domains. This protein localizes to focal adhesion sites and along actin stress fibers. Recruitment of this protein to the plasma membrane occurs in a lysophosphatidic acid (LPA)-dependent manner and it regulates LPA-induced cell migration. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC10556; MGC10558; MGC29959; MGC3837; MGC4423; OIP-1; OPA-interacting protein 1; tcag7.1300; thyroid hormone receptor interacting protein 6; Thyroid receptor-interacting protein 6; TR-interacting protein 6; ZRP-1; zyxin related protein 1; Zyxin-related protein 1
Gene Aliases: OIP-1; OIP1; TRIP-6; TRIP6; TRIP6i2; ZRP-1
UniProt ID: (Human) Q15654
Entrez Gene ID: (Human) 7205
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support