Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: KFSAACGPP VTPECEHCGQ RHQLGGPMWA EPIHDLDFVG RVLEAVSANP GRFHTSERIR GVLSVITEEL PDVPLYYTLD QLSSTIHCNT PSLLQLRSAL LHADFRVSLS HACKNAVKTD APASALWDIM RCWEKECPVK RERLSETSPA FRILSVEPRL QANFTIREDA NPSSRQRGLK RFQANPEANW GPRPRARPGG KAADEAMEER RRLLQNKRKE PPEDVAQRAA RLKTFPCKRF KEGTCQRGDQ CCYSHSPPTP RVSADAAPDC PETSNQTPPG PGAAAGPGID (371-659 aa encoded by BC002492)
TRMT1 is a tRNA(m(2)(2)G(26))dimethyltransferase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20244; N(2),N(2)-dimethylguanosine tRNA methyltransferase; TRM1 tRNA methyltransferase 1 homolog; tRNA (guanine(26)-N(2))-dimethyltransferase; tRNA 2,2-dimethylguanosine-26 methyltransferase; tRNA methyltransferase 1 homolog; tRNA(guanine-26,N(2)-N(2)) methyltransferase; tRNA(m(2,2)G26)dimethyltransferase
Gene Aliases: 6720406L13Rik; 6720477L24; D8Ertd812e; RGD1305992; TRM1; TRMT1
UniProt ID: (Human) Q9NXH9, (Mouse) Q3TX08
Entrez Gene ID: (Human) 55621, (Mouse) 212528, (Rat) 288914
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support