Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen Sequence: KLLQVGYTPD GKDDYRWCFR VDEVNWTTWN TNVGIINEDP GNCEGVKRTL SFSLRSSRVS GRHWKNFALV PLLREASARD RQSAQPEEVY LRQFSGSLKP EDAEVFKSPA ASGEK
TRPV1 (Transient Receptor Potential Cation Channel Subfamily V Member 1) is a receptor for capsaicin, the main pungent ingredient in hot chili peppers, and elicits a sensation of burning pain by selectively activating sensory neurons. TRPV1 is a non-selective cation channel that is structurally related to members of the TRP family of ion channels such as TRPC3 and TRPC6. TRPV1 is permeable to calcium, magnesium, and monovalent cations of sodium and potassium. Structurally, TRPV1 contains 6 trans-membrane domains, a P-loop membrane domain and 3 ankyrin repeat domains. TRPV1 is also activated by increases in temperature in the noxious range, suggesting that it functions as a transducer of painful thermal stimuli in vivo. TRPV1 seems to mediate proton influx and may be involved in intracellular acidosis in nociceptive neurons. The expression of TRPV1 appears to be limited to neurons within the sensory ganglia. Four transcript variants encoding TRPV1 with variations in th 5' UTR sequence have been described. Diseases associated with TRPV1 include Irritable Bowel Syndrome (IBS) and Bladder Disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Capsaicin receptor; cation channel; DKFZp434K0220; Osm-9-like TRP channel 1; OTRPC1; Transient receptor potential cation channel subfamily V member 1; transient receptor potential cation channel V1; transient receptor potential vanilloid 1 supraoptic nucleus; TrpV1; Vanilloid receptor; Vanilloid receptor 1; vanilloid receptor subtype 1; vanilloid receptor type 1 like protein 1; Vanilloid receptor type 1-like; vanilloid type 1 receptor
Gene Aliases: OTRPC1; Trpv1; TRPV1_SON; TRPV1alpha; TRPV1beta; VR-1; VR.5'sv; Vr1; Vr1l; Vr1l1
UniProt ID: (Rat) O35433, (Mouse) Q704Y3
Entrez Gene ID: (Rat) 83810, (Mouse) 193034
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support