Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TRPV4 Antibodies

          Osenses

          TRPV4 Polyclonal Antibody

          2 References
          View all (27) TRPV4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite TRPV4 Polyclonal Antibody

          Product Details

          OSR00136W-100UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:2,000
          -

          Immunohistochemistry (IHC)

          1:1,000-1:2,000
          -

          Miscellaneous PubMed (MISC)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / Ig

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide from aa region 300-400 of mouse TRPV4 conjugated to blue carrier protein was used as the antigen
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.

          The peptide is homologous in rat and human.

          Specificity of this antibody: TRPV4.

          Target Information

          TRPV4 (Transient Receptor Potential Cation Channel Subfamily V Member 4) is a cation-selective channel, activated in response to systemic osmotic pressure. Non-selective calcium permeant cation channel probably involved in osmotic sensitivity and mechanosensitivity. TRPV4 is a member of the OSM9-like transient receptor potential channel (OTRPC) subfamily in the transient receptor potential (TRP) superfamily of ion channels. The TRPV4 protein is a Ca2+-permeable, nonselective cation channel that is thought to be involved in the regulation of systemic osmotic pressure. Mutations in the TRPV4 gene are the cause of spondylometaphyseal and metatropic dysplasia and hereditary motor and sensory neuropathy type IIC. Diseases associated with TRPV4 include Metatropic Dysplasia and Spondylometaphyseal Dysplasia, Kozlowski Type. Multiple transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Osm-9-like TRP channel 4; OSM9-like transient receptor potential channel 4; osmosensitive transient receptor potential channel 4; OTRPC4; Transient receptor potential cation channel subfamily V member 4; transient receptor potential cation channel, subfamily 5, member 4; transient receptor potential cation channel, subfamily V, member 4; Transient receptor potential protein 12; TRP12; TrpV4; trpv4 channel; Vanilloi; Vanilloid receptor-like channel 2; Vanilloid receptor-like protein 2; vanilloid receptor-related osmotically activated channel; vanilloid receptor-related osmotically activated channel (Vroac); Vanilloid receptor-related osmotically-activated channel; VR-OAC; VRL-2

          View more View less

          Gene Aliases: 0610033B08Rik; BCYM3; CMT2C; HMSN2C; OTRPC4; SMAL; SPSMA; SSQTL1; TRP12; TRPV4; VR-OAC; VRL-2; VRL2; VROAC

          View more View less

          UniProt ID: (Human) Q9HBA0, (Mouse) Q9EPK8, (Rat) Q9ERZ8

          View more View less

          Entrez Gene ID: (Human) 59341, (Mouse) 63873, (Rat) 66026

          View more View less

          Function(s)
          actin binding osmosensor activity protein kinase C binding cation channel activity calcium channel activity protein binding calmodulin binding ATP binding microtubule binding stretch-activated, cation-selective, calcium channel activity protein kinase binding SH2 domain binding alpha-tubulin binding beta-tubulin binding actin filament binding nucleotide binding ion channel activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter diet induced thermogenesis calcium ion transport cellular calcium ion homeostasis cell volume homeostasis actin filament organization cell-cell junction assembly positive regulation of cytosolic calcium ion concentration osmosensory signaling pathway response to mechanical stimulus positive regulation of gene expression positive regulation of macrophage chemotaxis negative regulation of neuron projection development vasopressin secretion positive regulation of microtubule depolymerization actin cytoskeleton reorganization response to insulin cellular response to heat hyperosmotic salinity response glucose homeostasis positive regulation of vascular permeability cortical microtubule organization positive regulation of JNK cascade microtubule polymerization regulation of response to osmotic stress positive regulation of inflammatory response multicellular organismal water homeostasis positive regulation of ERK1 and ERK2 cascade calcium ion import calcium ion transmembrane transport cellular response to osmotic stress cellular hypotonic response cellular hypotonic salinity response positive regulation of monocyte chemotactic protein-1 production positive regulation of macrophage inflammatory protein 1 alpha production positive regulation of chemokine (C-C motif) ligand 5 production blood vessel endothelial cell delamination negative regulation of brown fat cell differentiation signal transduction involved in regulation of aerobic respiration positive regulation of chemokine (C-X-C motif) ligand 1 production positive regulation of energy homeostasis positive regulation of interleukin-6 secretion transport ion transport response to osmotic stress hypotonic response transmembrane transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0