Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: MGGFLPKAEG PGSQLQKLLP SFLVREQDWD QHLDKLHMLQ QKRILESPLL RASKENDLSV LRQLLLDCTC DVRQRGALGE TALHIAALYD NLEAALVLME
This gene is a member of the transient receptor family and the TrpV subfamily. The calcium-selective channel encoded by this gene has 6 transmembrane-spanning domains, multiple potential phosphorylation sites, an N-linked glycosylation site, and 5 ANK repeats. This protein forms homotetramers or heterotetramers and is activated by a low internal calcium level.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calciu; Calcium transport protein 2; calcium transporter 2; CaT2; ECaC; Epithelial calcium channel 1; Osm-9-like TRP channel 3; OTRPC3; Transient receptor potential cation channel subfamily V member 5; transient receptor potential cation channel, subfamily V, member 5; TrpV5
Gene Aliases: CAT2; ECAC1; OTRPC3; TRPV5
UniProt ID: (Human) Q9NQA5
Entrez Gene ID: (Human) 56302
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support