Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Calcium-permeable channels, such as TRPV6, participate in neurotransmission, muscle contraction, and exocytosis by providing calcium as an intracellular second messenger. Depending on the tissue, transcellular calcium transport may be regulated by vitamin D, parathyroid hormone (PTH), or calcitonin (CALCA). Calcium selective cation channel probably involved in Ca(2+) uptake in various tissues, including Ca(2+) reabsorption in intestine. The channel is activated by low internal calcium level, probably including intracellular calcium store depletion, and the current exhibits an inward rectification. Inactivation includes both, a rapid Ca(2+)-dependent and a slower Ca(2+)-calmodulin-dependent mechanism, the latter may be regulated by phosphorylation. In vitro, is slowly inhibited by Mg(2+) in a voltage-independent manner. Heteromeric assembly with TRPV5 seems to modify channel properties. TRPV5-TRPV6 heteromultimeric concatemers exhibit voltage-dependent gating. Interacts with TRPV5. Interacts with S100A10 and probably with the ANAX2-S100A10 heterotetramer. The interaction with S100A10 is required for the trafficking to the plasma membrane. Interacts with BSPRY. Interacts with calmodulin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alu-binding protein with zinc finger domain; Calcium transport protein 1; calcium transporter-like protein; CaT-L; CaT-like; CaT1; ECaC2; epithelial apical membrane calcium transporter/channel CaT1; Epithelial calcium channel 2; Transient receptor potential cation channel subfamily V member 6; TrpV6
Gene Aliases: ABP/ZF; CAT1; CATL; ECAC2; HSA277909; LP6728; TRPV6; ZFAB
UniProt ID: (Human) Q9H1D0
Entrez Gene ID: (Human) 55503
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support