Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The TSCOT gene, known as the Thymic Stromal Co-transporter, plays a crucial role in the gene expression regulation within thymic epithelial cells, essential for T cell development. It encodes a protein that bears weak homology to bacterial 12 transmembrane co-transporters. Research indicates that TSCOT is predominantly expressed in thymic tissues, although low levels of expression are detectable in other epithelial tissues such as skin and lungs. The gene has been co-analyzed with Foxn1 and Hoxa3, highlighting its significant role in organogenesis. Studies utilizing competitive reverse transcription-polymerase chain reaction (RT-PCR) have mapped TSCOT to specific genomic regions, verifying its expression in a tissue and cell-type specific manner. The introduction of a neoantigen into TSCOT-expressing cells demonstrates its capability to mediate antigen-specific T cell tolerance, offering potential applications in immunological therapies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: putative thymic stromal co-transporter TSCOT; Solute carrier family 46 member 2; solute carrier family 46, member 2; thymic stromal co-transporter; Thymic stromal cotransporter homolog; thymic stromal cotransporter protein
Gene Aliases: 5430429N04Rik; Ly110; SLC46A2; TSCOT; TSO-1C12
UniProt ID: (Human) Q9BY10, (Mouse) Q8CA03
Entrez Gene ID: (Human) 57864, (Mouse) 30936
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support