Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: SCVKYLIFLS NFPFSLLGLL ALAIGLWGLA VKGSLGSDLG GPLPADPMLG
Sequence homology: Human: 100%
TSPAN10 belongs to the tetraspanin (TM4SF) family. It is a multi-pass membrane protein. The exact function of TSPAN10 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Oculospanin; Tetraspanin-10; Tspan-10
Gene Aliases: OCSP; TSPAN10
UniProt ID: (Human) Q9H1Z9
Entrez Gene ID: (Human) 83882
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support