Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: GVWTYEQEL MVPVQWSDMV TLKARMTNYG LPRYRWLTHA WNFFQREFKC CGVVYFTDWL EMTEMDWPPD SCCVREFPGC SKQAHQEDLS DLYQEGCGKK MYSFLRGTKQ LQVLR (111-224 aa encoded by BC031265)
TSPAN12 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tetraspan NET-2; Tetraspanin-12; Transmembrane 4 superfamily member 12; Tspan-12
Gene Aliases: 9030619E17; AI426782; AI663988; AW111457; EVR5; NET-2; NET2; TM4SF12; TSPAN12; UNQ774/PRO1568
UniProt ID: (Human) O95859, (Rat) Q569A2, (Mouse) Q8BKT6
Entrez Gene ID: (Human) 23554, (Rat) 362326, (Mouse) 269831
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support