Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KSKSDRIVNE TLYENTKLLS ATGESEKQFQ EAIIVFQEEF KCCGLVNGAA DWGNNFQHYP ELCACLDKQR PCQSYNGKQV YKETCISFIK DFLAKN
TSPAN8 (tetraspanin 8), also known as TM4SF3 or CO-029, is a 237 amino acid multi-pass membrane protein that belongs to the tetraspanin family of transmembrane proteins and is thought to play a role in signal transduction events that influence cell development, activation, growth and motility. Localized to the cell membrane, TSPAN8 is expressed in colon, gastric, renal and pancreatic cancers, strongly suggesting a role in cancer formation and metastasis. The gene encoding TSPAN8 maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tetraspanin-8; Tetraspanin8; Transmembrane 4 superfamily member 3; Tspan-8; Tumor-associated antigen CO-029
Gene Aliases: CO-029; TM4SF3; TSPAN8
UniProt ID: (Human) P19075
Entrez Gene ID: (Human) 7103
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support