Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TSPAN8 Antibodies

          Invitrogen

          TSPAN8 Monoclonal Antibody (657909)

          1 Published Figure
          1 Reference
          View all (15) TSPAN8 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite TSPAN8 Monoclonal Antibody (657909)

          • Antibody Testing Data (2)
          • Published Figures (1)
          TSPAN8 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          TSPAN8 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TSPAN8 Antibody (MA5-24296) in IHC (F)

          Immunohistochemical analysis of TSPAN8 in perfusion fixed frozen sections of mouse intestine. Samples were incubated with TSPAN8 monoclonal antibody (Product # MA5-24296) using a dilution of 8 µg/mL overnight at 4 °C followed by NorthernLights™ 557-conjugated Anti-Rat IgG Secondary Antibody (yellow) and counterstained with DAPI (blue). Specific staining was localized to membranes of epithelial cells. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TSPAN8 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          TSPAN8 Antibody in Flow Cytometry (Flow)
          TSPAN8 Antibody in Immunohistochemistry (IHC)

          Product Details

          MA5-24296

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          8-25 µg/mL
          -

          Flow Cytometry (Flow)

          2.5 µg per million cells
          -
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Rat / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          657909

          Immunogen

          CHO Chinese hamster ovary cell line transfected with mouse TSPAN8

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.5 mg/mL

          Purification

          Protein A/G

          Storage buffer

          PBS with 5% trehalose

          Contains

          No Preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2606498

          Product Specific Information

          This antibody detects mouse TSPAN8 transfectants but not the parental cell line in flow cytometry.

          Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

          Target Information

          TSPAN8 (tetraspanin 8), also known as TM4SF3 or CO-029, is a 237 amino acid multi-pass membrane protein that belongs to the tetraspanin family of transmembrane proteins and is thought to play a role in signal transduction events that influence cell development, activation, growth and motility. Localized to the cell membrane, TSPAN8 is expressed in colon, gastric, renal and pancreatic cancers, strongly suggesting a role in cancer formation and metastasis. The gene encoding TSPAN8 maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Tetraspanin-8; Tetraspanin8; transmembrane 4 superfamily member 3; Tspan-8

          View more View less

          Gene Aliases: C76990; E330007O21Rik; Tm4sf3; Tspan8

          View more View less

          UniProt ID: (Mouse) Q8R3G9

          View more View less

          Entrez Gene ID: (Mouse) 216350

          View more View less

          Function(s)
          molecular_function integrin binding
          Process(es)
          negative regulation of blood coagulation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0