Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: IMLFFRSNPY FQNKVITKEY LVNITEYRAS HSTPIEWYPD YEVEAYRRRH HNSSLNFFNW FSDHNFAGSN KIAEILCKDL WRNPLQYYKR MKPPEEGTET SGDSQLLS
May be involved in sperm differentiation and proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 78; CT78; Testis-specific Y-encoded protein 1
Gene Aliases: CT78; DYS14; pJA923; TSPY; TSPY1
UniProt ID: (Human) Q01534
Entrez Gene ID: (Human) 7258
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support