Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: LKRKFIQMRR PFLERRDLII QHIPGFWVKA FLNHPRISIL INRRDEDIFR YLTNLQVQDL RHISMGYKMK LYFQTNPYFT NMVIVKEFQR NRSGRLVSHS TPIRWHRGQE PQARRHGNQD ASHSFFSWFS NHSLPEADRI AEIIKNDLWV NPLRYYLRER GSRIKRKKQE MKKRKTRGRC EVVIMEDAPD YYAVEDIFSE ISDIDETIHD IKISDFMETT D
This gene encodes the beta-subunit of glucosidase II, an N-linked glycan-processing enzyme in the endoplasmic reticulum (ER). This protein is an acidic phospho-protein known to be a substrate for protein kinase C. Mutations in this gene have been associated with the autosomal dominant polycystic liver disease (PCLD). Alternatively spliced transcript variants encoding distinct isoforms have been observed.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CASK interacting nucleosome assembly protein; CASK-interacting nucleosome assembly protein; Cell division autoantigen 1; cell division autoantigen 1 nucleolar protein; CTCL tumor antigen se20-4; CTCL-associated antigen se20-4; Cutaneous T-cell lymphoma-associated antigen se20-4; cutaneous T-cell lymphoma-associated tumor antigen se20-4; differentially expressed nucleolar TGF-beta1 target; Differentially-expressed nucleolar TGF-beta1 target protein; NP79; Nuclear protein of 79 kDa; nucleolar TGF-beta1 target protein; RP1-290F12.2; testis-specific protein Y encoded-like 2; Testis-specific Y-encoded-like protein 2; TSPY-like 2; TSPY-like protein 2
Gene Aliases: CDA1; CINAP; CTCL; DENTT; DXBwg1396e; DXHXS1008E; E130307F10Rik; HRIHFB2216; NP79; RGD1563283; SE204; TSPX; TSPYL2
UniProt ID: (Human) Q9H2G4, (Mouse) Q7TQI8
Entrez Gene ID: (Human) 64061, (Mouse) 52808, (Rat) 302612
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support