Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MALLGSRAE LEADEDVFED ALETISISSH SDMATSSLHF ASCDTQQAPR QRGASTVSSS SSTKVDLKSG LEECAVALNL FLSNKFTDAL ELLRPWAKES MYHALGYSTI VVLQAVLTFE QQDIQNGISA MKDALQTCQK KKYTVVESFS SLLSRGSLEQ LSEEEMHA (1-167 aa encoded by BC142985)
Regulates high density lipoprotein (HDL) cholesterol metabolism by promoting the ubiquitination and degradation of the oxysterols receptors LXR (NR1H2 and NR1H3). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tetratricopeptide repeat protein 39B; TPR repeat protein 39B
Gene Aliases: 1810054D07Rik; 9130422G05Rik; AI450603; AV103077; AW215585; C9orf52; T39; TTC39B
UniProt ID: (Human) Q5VTQ0, (Mouse) Q8BYY4
Entrez Gene ID: (Human) 158219, (Mouse) 69863
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support