Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive Samples: HeLa, Jurkat, HL-60, Mouse brain, Mouse spinal cord, Mouse heart, Rat brain
Immunogen sequence: MEAERGPERR PAERSSPGQT PEEGAQALAE FAALHGPALR ASGVPERYWG RLLHKLEHEV FDAGEVFGIM QVEEVEEEED EAAREVRKQQ PNPGNELCYK VIVTRESGLQ AAHPNSIFLI DHAWTCRVEH ARQQLQQVPG LLHRMANLMG IEFHGELPST EAVALVLEEM WKFNQTYQLA HGTAEEKMPV WYIMDEFGSR IQHADVPSFA TAPFFYMPQQ VAYTLLWPLR DLDTGEEVTR DFAYGETDPL IRKCMLLPWA PTDMLDLSSC TPEPPAEHYQ AILEENKEKL PLDINPVVHP HGHIFKVYTD VQQVASSLTH PRFTLTQSEA DADILFNFSH FKDYRKLSQE RPGVLLNQFP CENLLTVKDC LASIARRAGG PEGPPWLPRT FNLRTELPQF VSYFQQRERW GEDNHWICKP WNLARSLDTH VTKSLHSIIR HRESTPKVVS
TTLL12 (tubulin tyrosine ligase-like family, member 12) is a 644 amino acid protein that contains one TTL domain. Of the fourteen members of the TTLL family that modify tubulin, TTLL12 is the least characterized member. TTLL12 is highly expressed in a multitude of metastatic prostate cancer cell lines, therefore, it is considered a target for tumor therapy. Overexpression of TTLL12 is suggested to alter chromosome ploidy, whereas downregulation of TTLL12 influence several post-translational modifications of tubulin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inactive tubulin--tyrosine ligase-like protein 12; TTL domain protein; ttll12; tubulin tyrosine ligase-like family member 12; tubulin tyrosine ligase-like family, member 12; Tubulin--tyrosine ligase-like protein 12
Gene Aliases: BC055368; D430005B17; dJ526I14.2; KIAA0153; RGD1305319; TTLL12
UniProt ID: (Human) Q14166, (Mouse) Q3UDE2
Entrez Gene ID: (Human) 23170, (Rat) 300105, (Mouse) 223723
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support