Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: TEGDRNIWIV KPGAKSRGRG IMCMDHLEEM LKLVNGNPVV MKDGKWVVQK YIERPLLIFG TKFDLRQWFL VTDWNPLTVW FYRDSYIRFS TQPFSLKNLD K
TTLL3 gene ontology annotations related to this gene include protein polyglycylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HOTTL; Tubulin monoglycylase TTLL3; tubulin tyrosine ligase-like family member 3; tubulin tyrosine ligase-like family, member 3; Tubulin--tyrosine ligase-like protein 3
Gene Aliases: HOTTL; PRO0207; TTLL3
UniProt ID: (Human) Q9Y4R7
Entrez Gene ID: (Human) 26140
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support