Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: DGLEDCCSRD ENEEEEGDSE CSSLSAVSPS ESVAMISRSC MEILTKPLSN HEKVVRPALI YSLFPNVPPT IYFGTRDERV EKLPWEQRKL LRWKMSTVTP NIVKQTIGRS HFKISKRNDD WLGCWGHHMK SPSFRSI
Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Glutamylase which preferentially modifies beta-tubulin and non-tubulin proteins, such as NAP1L1, NAP1L4 and CGAS. Involved in the side-chain initiation step of the polyglutamylation reaction rather than in the elongation step. Involved in formation of short side-chains. Mediates initiation of polyglutamylation of nucleosome assembly proteins NAP1L1 and NAP1L4. Also acts as a monoglutamylase: generates monoglutamylation of CGAS, leading to impair the nucleotidyltransferase activity of CGAS. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein monoglutamylase TTLL4; Tubulin monoglutamylase TTLL4; tubulin tyrosine ligase-like family member 4; tubulin tyrosine ligase-like family, member 4; Tubulin--tyrosine ligase-like protein 4
Gene Aliases: KIAA0173; TTLL4
UniProt ID: (Human) Q14679
Entrez Gene ID: (Human) 9654
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support