Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Predicted to enable tubulin binding activity and tubulin-glutamic acid ligase activity. Predicted to be involved in microtubule cytoskeleton organization and protein polyglutamylation. Predicted to act upstream of or within microtubule bundle formation; microtubule severing; and positive regulation of cilium movement. Located in ciliary basal body.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein polyglutamylase TTLL6; TTLL6; Tubulin polyglutamylase TTLL6; tubulin tyrosine ligase-like family member 6; Tubulin--tyrosine ligase-like protein 6
Gene Aliases: TTL.6; TTLL6
UniProt ID: (Human) Q8N841
Entrez Gene ID: (Human) 284076
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support