Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TUBA3C Antibodies

          Abnova

          TUBA2 Monoclonal Antibody (3F10-2F2)

          View all (6) TUBA3C antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite TUBA2 Monoclonal Antibody (3F10-2F2)

          • Antibody Testing Data (1)
          TUBA2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TUBA2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TUBA2 Antibody (H00007278-M01) in WB

          TUBA2 monoclonal antibody (M01), clone 3F10-2F2. Western Blot analysis of TUBA2 expression in Jurkat. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TUBA2 Antibody in Western Blot (WB)

          Product Details

          H00007278-M01

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          3 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -

          ELISA (ELISA)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3F10-2F2

          Immunogen

          TUBA2 (AAH11721.1, 1 a.a. approximately 418 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.45 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          Immunogen sequence: MRECISIHVG QAGVQIGNAC WELYCLEHGI QPDGQMPSDK TIGGGDDSFN TFFSETGAGK HVPRAVFVDL EPTVVDEVRT GTYRQLFHPE QLITGKEDAA NNYARGHYTI GKEIVDLVLD RIRKLADLCT GLQGFLIFHS FGGGTGSGFA SLLMERLSVD YGKKSKLEFA IYPAPQVSTA VVEPYNSILT THTTLEHSDC AFMVDNEAIY DICRRNLDIE RPTYTNLNRL IGQIVSSITA SLRFDGALNV DLTEFQTNLV PYPRIHFPLA TYAPVISAEK AYHEQLSVAE ITNACFEPAN QMVKCDPRHG KYMACCMLYR GDVVPKDVNA AIATIKTKRT IQFVDWCPTG FKVGINYQPP TVVPGGDLAK VQRAVCMLSN TTAIAEAWAR LDLAALEKDY EEVGVDSVEA EAEEGEEY

          Target Information

          The microtubules are intracellular dynamic polymers made up of evolutionarily conserved polymorphic alpha/beta-tubulin heterodimers and a large number of microtubule-associated proteins (MAPs). Microtubules have their intrinsic polarity; highly dynamic plus ends and less dynamic minus ends. Microtubules are required for vital processes in eukaryotic cells including mitosis, meiosis, maintenance of cell shape and intracellular transport. Microtubules are also necessary for movement of cells by means of flagella and cilia. In mammalian tissue culture cells microtubules have their minus ends anchored in microtubule organizing centers (MTOCs).The GTP (guanosintriphosphate) molecule is an essential for tubulin heterodimer to associate with other heterodimers to form microtubule. In vivo, microtubule dynamics vary considerably. Microtubule polymerization is reversible and a populations of microtubules in cells are on their minus ends either growing or shortening - this phenomenon is called dynamic instability of microtubules. The alpha-tubulin (relative molecular weight around 50 kDa) is globular protein that exists in cells as part of soluble alpha/beta-tubulin dimer or it is polymerized into microtubules. In different species it is coded by multiple tubulin genes that form tubulin classes (in human 6 genes).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alpha tubulin 3C; Alpha-tubulin 2; Alpha-tubulin 3C; alpha-tubulin 3C/D; Tubulin alpha-2 chain; Tubulin alpha-3C chain; tubulin, alpha 2; tubulin, alpha 3c

          View more View less

          Gene Aliases: bA408E5.3; TUBA2; TUBA3C

          View more View less

          UniProt ID: (Human) P0DPH7

          View more View less

          Entrez Gene ID: (Human) 7278

          View more View less

          Function(s)
          GTPase activity structural constituent of cytoskeleton protein binding GTP binding
          Process(es)
          cytoskeleton organization microtubule-based process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-79b49c4fc5-ghzkj:80/100.66.129.252:80.
          git-commit: cca8d8ff64ba87988ed84f3fa7b2b2bee312c353
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0