Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MNGTRNWCT LVDVHPEDQA AAGRKTYAMV SSHSAGHSLA SELVESHDGH EEIIKVYLKG RSGDKMIHEK NINQLKSEVQ YIQEARNCLQ KLREDISSKL DRNLGDSLHR QEIQVVLEKP NGFSQSPTAL YSSPPEVDTC INEDVESLRK TVQDLLAKLQ EAKRQHQSDC VAFEVTLSRY QREAEQSNVA LQREEDRVEQ KEAEVGELQR RLLGMETEHQ ALLAKVREGE VALEELRSNN ADCQAEREKA ATLEKEVAGL REKIHHLDDM LKSQQRKVRQ MIEQLQNSKA VIQSKDATIQ ELKEKIAYLE AENLEMHDRM EHLIEKQISH GNFSTQARAK TENPGSIRIS KPPSPKPMPV IRVVET (1-365 aa encoded by B C002933)
TUFT1 is involved in the mineralization and structural organization of enamel.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tuftelin; UNQ865
Gene Aliases: TUFT1
UniProt ID: (Human) Q9NNX1
Entrez Gene ID: (Human) 7286
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support