Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MSQDNDTLMR DILGHELAAM RLQKLEQQRR LFEKKQRQKR QELLMVQANP DASPWLWRSC LREERLLGDR GLGNPFLRKK VSEAHLPSGI HSALGTVSCG GDGRGERGLP TPRTEAVFRN LGLQSPFLSW LPDNSDAELE EVSVENGSVS PPPFKQSPRI RRKGWQAHQR PGTRAEGESD SQDMGDAHKS PNMGPNPGMD GDCVYENLAF QKEEDLEKKR EASESTGTNS SAAHNEELSK ALKGEGGTDS DHMRHEASLA IRSPCPGLEE DMEAYVLRPA LPGTMMQCYL TRDKHGVDKG LFPLYYLYLE TSDSLQRFLL AGRKRRRSKT SNYLISLDPT HLSRDGDNFV GKVRSNVFST KFTIFDNGVN PDREHLTRNT ARIRQELGAV CYEPNVLGYL GPRKMTVILP GTNSQNQRIN VQPLNEQESL LSRYQRGDKQ GLLLLHNKTP SWDKENGVYT LNFHGRVTRA SVKNFQIVDP KHQEHLVLQF GRVGPDTFTM DFCFPFSPLQ AFSICLSSFN
TULP2 is a member of a family of tubby-like genes (TULPs) that encode proteins of unknown function. Members of this family have been identified in plants, vertebrates, and invertebrates. The TULP proteins share a conserved C-terminal region of approximately 200 amino acid residues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer testis antigen 65; Cancer/testis antigen 65; CT65; Tubby-like protein 2; Tubby-related protein 2
Gene Aliases: CT65; TUBL2; TULP2
UniProt ID: (Human) O00295
Entrez Gene ID: (Human) 7288
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support