Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Sequence of this protein is as follows: MASKKREVQL QTVINNQSLW DEMLQNKGLT VIDVYQAWCG PCRAMQPLFR KLKNELNEDE ILHFAVAEAD NIVTLQPFRD KCEPVFLFSV NGKIIEKIQG ANAPLVNKKV INLIDEERKI AAGEMARPQY PEIPLVDSDS EVSEESPCES VQELYSIAII KPDAVISKKV LEIKRKITKA GFIIEAEHKT VLTEEQVVNF YSRIADQRDF EEFVSFMTSG LSYILVVSQG SKHNPPSEET EPQTDTEPNE RSEDQPEVEA QVTPGMMKNK QDSLQEYLER QHLAQLCDIE EDAANVAKFM DAFFPDFKKM KSMKLEKTLA LLRPNLFHER KDDVLRIIKD EDFKILEQRQ VVLSEKEAQA LCKEYENEDY FNKLIENMTS GPSLALVLLR DNGLQYWKQL LGPRTVEEAI EYFPESLCAQ FAMDSLPVNQ LYGSDSLETA EREIQHFFPL QSTLGLIKPH ATSEQREQIL KIVKEAGFDL TQVKKMFLTP EQTEKIYPKV TGKDFYKDLL EMLSVGPSMV MILTKWNAVA EWRRLMGPTD PEEAKLLSPD SIRAQFGISK LKNIVHGASN AYEAKEVVNR LFEDPEEN
This gene encodes a protein with an N-terminal thioredoxin domain and three C-terminal nucleoside diphosphate kinase (NDK) domains, but the NDK domains are thought to be catalytically inactive. The sea urchin ortholog of this gene encodes a component of sperm outer dynein arms, and the protein is implicated in ciliary function. Mutations in this gene are implicated in primary ciliary dyskinesia type 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'-5' exonuclease NME8; epididymis secretory protein Li 99; NM23-H8; NME/NM23 family member 8; sperm-specific thioredoxin 2; Spermatid-specific thioredoxin-2; Sptrx-2; thioredoxin domain containing 3 (spermatozoa); Thioredoxin domain-containing protein 3
Gene Aliases: CILD6; HEL-S-99; NM23-H8; NME8; sptrx-2; SPTRX2; TXNDC3
UniProt ID: (Human) Q8N427
Entrez Gene ID: (Human) 51314
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support