Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MHPAVFLSLP DLRCSLLLLV TWVFTPVTTE ITSLDTENID EILNNADVAL VNFYADWCRF SQMLHPIFEE ASDVIKEEFP NENQVVFARV DCDQHSDIAQ RYRISKYPTL KLFRNGMMMK REYRGQRSVK ALADYIRQQK SDPIQEIRDL AEITTLDRSK RNIIGYFEQK DSDNYRVFER VANILHDDCA FLSAFGDVSK PERYSGDNII YKPPGHSAPD MVYLGAMTNF DVTYNWIQDK CVPLVREITF ENGEELTEEG LPFLILFHMK EDTESLEIFQ NEVARQLISE KGTINFLHAD CDKFRHPLLH IQKTPADCPV IAIDSFRHMY VFGDFKDVLI PGKLKQFVFD LHSGKLHREF HHGPDPTDTA PGEQAQDVAS SPPESSFQKL APSEYRYTLL RDRDEL
Mediates thiol-dependent retention in the early secretory pathway, forming mixed disulfides with substrate proteins through its conserved CRFS motif. Inhibits the calcium channel activity of ITPR1. May have a role in the control of oxidative protein folding in the endoplasmic reticulum. Required to retain ERO1L and ERO1LB in the endoplasmic reticulum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endoplasmic reticulum resident protein 44; endoplasmic reticulum resident protein 44 kDa; endoplasmic reticulum resident protein 44kDa; endoplasmic reticulum resident protein ERp44; ER protein 44; OTTHUMP00000063799; protein disulfide isomerase family A, member 10; thioredoxin domain containing 4 (endoplasmic reticulum); Thioredoxin domain-containing protein 4
Gene Aliases: 1110001E24Rik; AI849526; AL033348; ERP44; KIAA0573; PDIA10; TXNDC4; UNQ532/PRO1075
UniProt ID: (Human) Q9BS26, (Mouse) Q9D1Q6
Entrez Gene ID: (Human) 23071, (Mouse) 76299
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support