Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: AAAAAADGPP AADGEDGQDP HSKHLYTADM FTHGIQSAAH FVMFFAPWCG HCQRLQPTWN DLGDKYNSME DAKVYVAKVD CTAHSDVCSA QGVRGYPTLK LFKPGQEAVK YQGPRDFQTL ENWMLQTLNE EPVTPEPEVE PPSAPELKQG LYELSASNFE LHVAQGDHFI KFFAPWCGHC KALAPTWEQL ALGLEHSETV KIGKVDCTQH YELCSGNQVR GYPTLLWFRD GKKVDQYKGK RDLESLREYV
This gene encodes a protein-disulfide isomerase. Its expression is induced by hypoxia and its role may be to protect hypoxic cells from apoptosis. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: endoplasmic reticulum protein ERp46; Endoplasmic reticulum resident protein 46; endothelial protein disulphide isomerase; ER protein 46; FLJ21353; FLJ90810; MGC3178; protein disulfide isomerase family A, member 15; thiore; thioredoxin domain containing 5 (endoplasmic reticulum); Thioredoxin domain-containing protein 5; thioredoxin related protein; Thioredoxin-like protein p46
Gene Aliases: ENDOPDI; ERP46; HCC-2; HCC2; PDIA15; STRF8; TLP46; TXNDC5; UNQ364; UNQ364/PRO700
UniProt ID: (Human) Q8NBS9
Entrez Gene ID: (Human) 81567
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support