Search Thermo Fisher Scientific
Compatible with Direct ELISA
Theralizumab is a monoclonal antibody that acts as a superagonist of the CD28 receptor on T cells, leading to their activation and proliferation.
Protein Aliases: CD28; TGN1412