Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Thickveins Antibodies

          Fabgennix

          Thickveins Polyclonal Antibody

          View all (3) Thickveins antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Thickveins Polyclonal Antibody

          • Antibody Testing Data (1)
          Thickveins Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Thickveins Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Thickveins Antibody (TKV-101AP) in WB

          Western blot of Thickveins antibody (TKV-101AP) with fruity fly extract. 1:500 antibody dilution in DiluOBuffer™ (FGI-1963). Rabbit # 623.pb1.AP Project # 1086B. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Thickveins Antibody in Western Blot (WB)

          Product Details

          TKV-101AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          ELISA (ELISA)

          1:10,000
          -

          Immunoprecipitation (IP)

          1:50-1:250
          -
          Product Specifications

          Species Reactivity

          Fruit fly

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to unique amino acid sequences on pan Thickvein protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          proprietary buffer, pH 7.4-7.8, with 0.5% BSA, 30% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Target Information

          Mutants of the thick veins (tkv) gene have been described previously (as slater alleles) as embryonic lethal, lacking dorsal epidermis, but not as showing a recognizable dorsoventral phenotype. The maternal alteration of function coupled with zygotic reduction of function of tkv is strongly ventralizing. In double heterozygous combinations in the mother, tkv mutations increase the ventralizing effect of dominant, weakly ventralizing alleles of the maternal effect, dorsoventral genes easter and cactus.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Activin-A-receptor-25D; BMP receptor kinase 25D; CG14026-PA; CG14026-PB; CG14026-PC; CG14026-PD; DPP receptor; Receptor protein serine/threonine kinase; slater; thick veins; Thick-vein; Thick-veins; thickvein; thickveins; tkv; tkv-PA; tkv-PB; tkv-PC; tkv-PD

          View more View less

          Gene Aliases: Atkv; Atr25D; Brk25D; Brk25D1; Brk25D2; CG14026; Dmel\CG14026; Dmel_CG14026; Dtfr; kv; l(2)04415; l(2)25Da; STK-A; str; TKV; TKV1; tkva

          View more View less

          Entrez Gene ID: (Fruit fly) 33753

          View more View less

          Function(s)
          protein kinase activity receptor signaling protein serine/threonine kinase activity transforming growth factor beta receptor activity, type I protein binding ATP binding transforming growth factor beta binding
          Process(es)
          compound eye morphogenesis morphogenesis of a branching structure regulation of transcription from RNA polymerase II promoter protein phosphorylation transforming growth factor beta receptor signaling pathway transforming growth factor beta receptor complex assembly neuromuscular synaptic transmission chorion-containing eggshell formation dorsal closure open tracheal system development anterior/posterior pattern specification, imaginal disc imaginal disc-derived wing morphogenesis histoblast morphogenesis heart development decapentaplegic signaling pathway germ cell migration regulation of synaptic growth at neuromuscular junction dorsal/ventral pattern formation negative regulation of gene expression cell migration regulation of cell-cell adhesion BMP signaling pathway ovarian follicle cell development germ-line stem cell population maintenance spectrosome organization genital disc development germ-line stem cell division regulation of imaginal disc growth regulation of cell differentiation negative regulation of salivary gland boundary specification positive regulation of synaptic growth at neuromuscular junction positive regulation of growth branched duct epithelial cell fate determination, open tracheal system wing disc anterior/posterior pattern formation positive regulation of muscle organ development transforming growth factor beta receptor signaling pathway involved in endodermal cell fate specification anterior Malpighian tubule development cell elongation involved in imaginal disc-derived wing morphogenesis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0