Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEGGGGSGNK TTGGLAGFFG AGGAGYSHAD LAGVPLTGMN PLSPYLNVDP RYLVQDTDEF IPTGANKTRG RFELAFFTIG GCCMTGAAFG AMNGLRLGLK ETQNMAWSKP RNVQILNMVT RQGALWANTL GSLALLYSAF GVIIEKTRGA EDDLNTVAAG TMTGMLYKCT GGLRGIARGG LTGLTLTSLY ALYNNWEHMK GSLLQQSL
Essential component of the TIM23 complex, a complex that mediates the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial import inner membrane translocase subunit Tim23; translocase of inner mitochondrial membrane 23 homolog; translocase of the inner mitochondrial membrane
Gene Aliases: TIM23; TIMM23
UniProt ID: (Human) O14925, (Rat) O35093
Entrez Gene ID: (Human) 100287932, (Rat) 54312, (Mouse) 53600
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support