Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Exalpha
Weakly expressed in kidney. Expressed in various tumor cell lines including carcinomas, myeloid and lymphoid malignancies, melanomas and prostate cancer. Overexpressed in taxol-resistant breast cancer line MDA 435(TR) and the doxorubicin-resistant multiple myelanoma lines 8226/Dox(40) and 8226/MDR(10)V.1.
The expression of the cancer-testis antigen Taxol resistanceassociated gene-3 (TRAG-3) is associated with acquired paclitaxel (Taxol) resistance, and is expressed in a variety of cancers - e.g., breast cancer, leukemia, and melanoma. Overexpressed in taxolresistant breast cancer line MDA 435(TR) and the doxorubicinresistant multiple myelanoma lines 8226/Dox(40) and 8226/MDR (10)V. Weakly expressed in kidney. TRAG-3 is an attractive target for immunotherapy of cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 24.2; cancer/testis antigen family 24, member 2; chondrosarcoma-associated gene 2/3 protein-like; CSAG family, member 3B; MGC149851; MGC149852; taxol resistance associated gene 3; taxol-resistant-associated gene 3 protein; trag 3
Gene Aliases: CSAG3B; CT24.2; TRAG-3; TRAG3
Entrez Gene ID: (Human) 102723547
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support