Search Thermo Fisher Scientific
The PA1-73101 antibody reacts with Treponema pallidum.
PA1-73101 has been successfully used in Immunofluorescence applications.
The PA1-73101 antibody was raised against Highly purified T. pallidum.
This antibody reacts with ultrasonicated lysates of T. pallidum and T. reiter.
Protein Aliases: Syphillis; t pallidum; T. pallidum