Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: IPATALLPT MTPDGLAVTP TPVPVVGSQM TRQARRLYVG NIPFGITEEA MMDFFNAQMR LGGLTQAPGN PVLAVQINQD KNFAFLEFRS VDETTQAMAF DGIIFQGQSL KIRRPHDYQP LPGMSENPSV YVPGVVSTVV PDSAHKLFIG GLPNYLNDDQ VKELLTSFGP LKAFNLVKDS ATGLSKGYAF CEYVDINVTD QAIAGLNGMQ LGDKKLLVQR ASVGAKNATL STINQTPVTL QVPGLMSSQV QMGGHPTEVL CLMNMVLPEE LLDDEEYEEI VEDVRDECSK YGLVKSIEIP RPVDGVEVPG CGKIFVEFTS VFDCQKAMQG LTGRKFANRV VVTKYCDPDS YHRRDFW (116-471 aa encoded by BC008740 )
U2 auxiliary factor (U2AF), comprised of a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the U2AF large subunit which contains a sequence-specific RNA-binding region with 3 RNA recognition motifs and an Arg/Ser-rich domain necessary for splicing. The large subunit binds to the polypyrimidine tract of introns early during spliceosome assembly. Multiple transcript variants have been detected for this gene, but the full-length natures of only two have been determined to date.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hU2AF(65); hU2AF65; Splicing factor U2AF 65 kDa subunit; U2 (RNU2) small nuclear RNA auxiliary factor 2; U2 auxiliary factor 65 kDa subunit; U2 small nuclear ribonucleoprotein auxiliary factor (65kD); U2 snRNP auxiliary factor large subunit
Gene Aliases: U2AF2; U2AF65
UniProt ID: (Human) P26368
Entrez Gene ID: (Human) 11338
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support