Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: AIHWSLPRAL VPATGTRASG SQKRQRPPKG QEALWSRPHS RHSAERAGGS
Sequence homology: Human: 100%
The protein encoded by this gene is a type II transmembrane protein that is similar to two families of cell adhesion molecules, the protocadherins and ependymins. This protein may play a role in calcium-dependent cell adhesion. This protein is glycosylated, and the orthologous mouse protein is localized to the lysosome. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 8.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ependymin related protein 1; mammalian ependymin related protein 1; Mammalian ependymin-related protein 1; MERP-1; Upregulated in colorectal cancer gene 1 protein
Gene Aliases: EPDR; EPDR1; MERP-1; MERP1; UCC1
UniProt ID: (Human) Q9UM22
Entrez Gene ID: (Human) 54749
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support