Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • UFSP2 Antibodies

          Proteintech

          UFSP2 Polyclonal Antibody

          1 Reference
          View all (4) UFSP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite UFSP2 Polyclonal Antibody

          • Antibody Testing Data (22)
          UFSP2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          UFSP2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          UFSP2 Antibody (16999-1-AP) in ICC/IF

          Immunofluorescent analysis of HeLa cells using 16999-1-AP (UFSP2 antibody) at dilution of 1:25 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          UFSP2 Antibody in Immunocytochemistry (ICC/IF)
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UFSP2 Antibody in Western Blot (WB)
          UFSP2 Antibody in Western Blot (WB)
          UFSP2 Antibody in Western Blot (WB)
          UFSP2 Antibody in Western Blot (WB)
          UFSP2 Antibody in Immunoprecipitation (IP)

          Product Details

          16999-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          UFSP2 Fusion Protein Ag10692 (120-469 aa encoded by BC010493)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.19 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          The MW of this protein is 53 kDa, and this antibody specifically recognizes the 53 kDa protein.

          Immunogen sequence: MLEMSTSLA AVTPIIERES GGHHYVNMTL PVDAVISVAP EETWGKVRKL LVDAIHNQLT DMEKCILKYM KGTSIVVPEP LHFLLPGKKN LVTISYPSGI PDGQLQAYRK ELHDLFNLPH DRPYFKRSNA YHFPDEPYKD GYIRNPHTYL NPPNMETGMI YVVQGIYGYH HYMQDRIDDN GWGCAYRSLQ TICSWFKHQG YTERSIPTHR EIQQALVDAG DKPATFVGSR QWIGSIEVQL VLNQLIGITS KILFVSQGSE IASQGRELAN HFQSEGTPVM IGGGVLAHTI LGVAWNEITG QIKFLILDPH YTGAEDLQVI LEKGWCGWKG PDFWNKDAYY NLCLPQRPNM I (120-469 aa encoded by BC010493)

          Target Information

          Ubiquitin-fold modifier 1 (Ufm1)-specific protease 2 (UfSP2) is a cysteine protease that is responsible for the release of Ufm1 from Ufm1-conjugated cellular proteins, as well as for the generation of mature Ufm1 from its precursor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: UFM1-specific peptidase 2; Ufm1-specific protease 2; UfSP2

          View more View less

          Gene Aliases: 1810047C23Rik; BHD; C4orf20; RGD1311161; UFSP2

          View more View less

          UniProt ID: (Human) Q9NUQ7, (Rat) Q5XIB4, (Mouse) Q99K23

          View more View less

          Entrez Gene ID: (Human) 55325, (Rat) 361151, (Mouse) 192169

          View more View less

          Function(s)
          protein binding thiolester hydrolase activity UFM1 hydrolase activity peptidase activity cysteine-type peptidase activity hydrolase activity
          Process(es)
          proteolysis regulation of intracellular estrogen receptor signaling pathway
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-2w6sl:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0