Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: LSYVPNDLLG FSDAMTFKER VWNHIVHLED HLFCQYLFRN ALEIASEILQ TPVTAYDLYS HTSIWLLRTD FVLDYPKPVM PNMIFIGGIN CHQGKPLPME FEA
This gene encodes a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UDP glucuronosyltransferase 1 family, polypeptide A10; UDP glycosyltransferase 1 family, polypeptide A10; UDP-glucuronosyltransferase 1-10; UDP-glucuronosyltransferase 1-J; UDP-glucuronosyltransferase 1A10; UDPGT 1-10; UGT-1J; UGT1A10
Gene Aliases: GNT1; UDPGT; UGT-1J; UGT1; UGT1-10; UGT1.10; UGT1A10; UGT1J
UniProt ID: (Human) Q9HAW8
Entrez Gene ID: (Human) 54575
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support