Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MENFILYEEI GRGSKTVVYK GRRKGTINFV AILCTDKCKR PEITNWVRLT REIKHKNIVT FHEWYETSNH LWLVVELCTG GSLKTVIAQD ENLPEDVVRE FGIDLISGLH HLHKLGILFC DISPRKILLE GPGTLKFSNF CLAKVEGENL EEFFALVAAE EGGGDNGENV LKKSMKSRVK GSPVYTAPEV VRGADFSISS DLWSLGCLLY EMFSGKPPFF SESISELTEK ILCEDPLPPI PKDSSRPKAS SDFINLLDGL LQRDPQKRLT WTRLLQHSFW KKAFAGADQE
The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. ULK4 (unc-51-like kinase 4) is a 1,275 amino acid protein that contains one protein kinase domain and one HEAT repeat and belongs to the Ser/Thr protein kinase family. Although containing what is thought to be a catalytically inactive domain, ULK4 may play a role in the ATP-dependent phosphorylation of target proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Serine/threonine-protein kinase ULK4; unc-51 like kinase 4; Unc-51-like kinase 4
Gene Aliases: 4932415A06Rik; A730098P15; FAM7C1; REC01035; RGD1565726; ULK4
UniProt ID: (Human) Q96C45, (Mouse) Q3V129
Entrez Gene ID: (Human) 54986, (Mouse) 209012, (Rat) 363167
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support