Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ULK4 Antibodies

          Abnova

          ULK4 Monoclonal Antibody (4A10-1A7)

          View all (13) ULK4 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ULK4 Monoclonal Antibody (4A10-1A7)

          • Antibody Testing Data (1)
          ULK4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ULK4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ULK4 Antibody (H00054986-M01) in WB

          Western Blot analysis of ULK4 expression in transfected 293T cell line by ULK4 monoclonal antibody (M01), clone 4A10-1A7. Lane 1: ULK4 transfected lysate(65 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ULK4 Antibody in Western Blot (WB)

          Product Details

          H00054986-M01

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          ELISA (ELISA)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          4A10-1A7

          Immunogen

          ULK4 (AAH14794, 1 a.a. approximately 580 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          Immunogen sequence: MENFILYEEI GRGSKTVVYK GRRKGTINFV AILCTDKCKR PEITNWVRLT REIKHKNIVT FHEWYETSNH LWLVVELCTG GSLKTVIAQD ENLPEDVVRE FGIDLISGLH HLHKLGILFC DISPRKILLE GPGTLKFSNF CLAKVEGENL EEFFALVAAE EGGGDNGENV LKKSMKSRVK GSPVYTAPEV VRGADFSISS DLWSLGCLLY EMFSGKPPFF SESISELTEK ILCEDPLPPI PKDSSRPKAS SDFINLLDGL LQRDPQKRLT WTRLLQHSFW KKAFAGADQE SSVEDLSLSR NTMECSGPQD SKELLQNSQS RQAKGHKSGQ PLGHSFRLEN PTEFRPKSTL EGQLNESMFL LSSRPTPRTS TAVEVSPGED MTHCSPQKTS PLTKITSGHL SQQDLESQMR ELIYTDSDLV VTPIIDNPKI MKQPPVKFDA KILHLPTYSV DKLLFLKDQD WNDFLQQVCS QIDSTEKSMG ASRAKLNLLC YLCVVAGHQE VVTRLLHSPL FQLLIQHLRI APNWDIRAKV AHVIGLLASH TAELQENTPV VETTSSIGIG ILNCLVQHST PVPRQCLVYV

          Target Information

          The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. ULK4 (unc-51-like kinase 4) is a 1,275 amino acid protein that contains one protein kinase domain and one HEAT repeat and belongs to the Ser/Thr protein kinase family. Although containing what is thought to be a catalytically inactive domain, ULK4 may play a role in the ATP-dependent phosphorylation of target proteins.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Serine/threonine-protein kinase ULK4; Unc-51-like kinase 4

          View more View less

          Gene Aliases: FAM7C1; REC01035; ULK4

          View more View less

          UniProt ID: (Human) Q96C45

          View more View less

          Entrez Gene ID: (Human) 54986

          View more View less

          Function(s)
          protein serine/threonine kinase activity ATP binding
          Process(es)
          microtubule cytoskeleton organization protein phosphorylation regulation of neuron projection development regulation of MAPK cascade regulation of JNK cascade regulation of protein kinase C signaling regulation of p38MAPK cascade regulation of neuron migration
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-9267x:80/100.66.134.224:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0