Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Rockland
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This protein A purified antibody is directed against human UPLC1/ASAP3 protein.A BLAST analysis was used to suggest cross-reactivity with UPLC1/ASAP3 protein from mouse and rat based on approximately 80% homology with the human protein. Reactivity against homologues from other sources is not known.
This gene encodes a member of a subfamily of ADP-ribosylation factor(Arf) GTPase-activating proteins that contain additional ankyrin repeat and pleckstrin homology domains. The Arf GAP domain of this protein catalyzes the hydrolysis of GTP bound to Arf proteins. The encoded protein promotes cell differentiation and migration and has been implicated in cancer cell invasion. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 3; ARF6 GTPase-activating protein; ArfGAP with SH3 domain, ankyrin repeat and PH domain 3 1; centaurin, beta 6; development and differentiation enhancing factor-like 1; Development and differentiation-enhancing factor-like 1; Protein up-regulated in liver cancer 1; up-regulated in liver cancer 1 (UPLC1)
Gene Aliases: ACAP4; ASAP3; CENTB6; DDEFL1; UPLC1
UniProt ID: (Human) Q8TDY4
Entrez Gene ID: (Human) 55616
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support