Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MDMLDPGLDP AASATAAAAA SHDKGPEAEE GVELQEGGDG PGAEEQTAVA ITSVQQAAFG DHNIQYQFRT ETNGGQVTYR VVQVTDGQLD GQGDTAGAVS VVSTAAFAGG QQAVTQVGVD GAAQRPGPAA ASVPPGPAAP FPLAVIQNPF SNGGSPAAEA VSGEARFAYF PASSVGDTTA VSVQTTDQSL QAGGQFYVMM TPQDVLQTGT QRTIAPRTHP YSPKIDGTRT PRDERRRAQH NEVERRRRDK INNWIVQLSK IIPDCNADNS KTGASKGGIL SKACDYIREL RQTNQRMQET FKEAERLQMD NELLRQQIEE LKNENALLRA QLQQHNLEMV GEGTRQ
USF2 consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHb12; c-fos interacting; c-fos interacting protein; Class B basic helix-loop-helix protein 12; FIP; FOS-interacting protein; Major late transcription factor 2; Upstream stimulatory factor 2; Upstream transcription factor 2
Gene Aliases: BHLHB12; FIP; USF2
UniProt ID: (Human) Q15853
Entrez Gene ID: (Human) 7392
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support