Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALHSPQYIF GDFSPDEFNQ FFVTPRSSVE LPPYSGTVLC GTQAVDKLPD GQEYQRIEFG VDEVIEPSDT LPRTPSYSIS STLNPQAPEF ILGCTASKIT PDGITKEASY GSIDCQYPGS ALALDGSSNV EAEVLENDGV SGGLGQRERK KKKKRPPGYY SYLKDGGDDS ISTEALVNGH ANSAVPNSVS AEDAEFMGDM PPPLTPRTCN SPQNSTDSVS DIVPDSPFPG ALGSDTRTAG QPEGGPGADF GQSCFPAEAG RDTLSRTAGA QPCVGTDTTE NLGVANGQIL ESSGEGTATN GVELHTTESI DLDPTKPESA SPPADGTGSA SGTLPVSQPK SWASLFHDSK PSSSSPVAYV ETKYSPPAIS PLVSEKQVEV KEGLVPVSED PVAIKIAELL ENVTLIHKPV SLQPRGLINK GNWCYINATL QALVACPPMY HLMKFIPLYS KVQRPCTSTP MIDSFVRLMN EFTNMPVPPK PRQALGDKIV RDIRPGAAFE PTYIYRLLTV NKSSLSEKGR QEDAEEYLGF ILNGLHEEML NLKKLLSPSN EKLTISNGPK NHSVNEEEQE EQGEGSEDEW EQVGPRNKTS VTRQADFVQT PITGIFGGHI RSVVYQQSSK ESATLQPFFT LQLDIQSDKI RTVQDALESL VARESVQGYT TKTKQEVEIS RRVTLEKLPP VLVLHLKRFV YEKTGGCQKL IKNIEYPVDL EISKELLSPG VKNKNFKCHR TYRLFAVVYH HGNSATGGHY TTDVFQIGLN GWLRIDDQTV KVINQYQVVK PTAERTAYLL YYRRVDLL
USP10, also known as ubiquitin specific peptidase 10, belongs to the ubiquitin-specific protease family of cysteine proteases. USP10 functions to catalyze the cleavage of ubiquitin from ubiquitin-conjugated protein substrates such as p53/TP53, SNX3 and CFTR. USP10 has been identified as a subunit of DNA-bound androgen receptor (AR) complexes and may play a role in the activity of the DNA-bound androgen receptor complex. USP10 also acts as an essential regulator of p53/TP53 stability and is thought to function as a tumor suppressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 10; MGC2621; Ubiquitin carboxyl-terminal hydrolase 10; ubiquitin specific protease 10; Ubiquitin thioesterase 10; ubiquitin thiolesterase 10; Ubiquitin-specific-processing protease 10
Gene Aliases: KIAA0190; UBPO; USP10
UniProt ID: (Human) Q14694
Entrez Gene ID: (Human) 9100
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support