Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • USP2 Antibodies

          Proteintech

          USP2 Polyclonal Antibody

          17 References
          View all (10) USP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite USP2 Polyclonal Antibody

          • Antibody Testing Data (18)
          USP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          USP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 18

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          USP2 Antibody (10392-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded Human breast cancer tissue slide using (Product # 10392-1-AP) USP2 antibody at dilution of 1:200 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          USP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP2 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          USP2 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          USP2 Antibody in Western Blot (WB)
          USP2 Antibody in Western Blot (WB)
          USP2 Antibody in Western Blot (WB)
          USP2 Antibody in Western Blot (WB)
          USP2 Antibody in Western Blot (WB)
          USP2 Antibody in Western Blot (WB)
          USP2 Antibody in Western Blot (WB)
          USP2 Antibody in Western Blot (WB)
          USP2 Antibody in Western Blot (WB)

          Product Details

          10392-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 10 publications 10 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          USP2 Fusion Protein Ag0591 (11-241 aa encoded by BC002955)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody is specific to the isoform 1 (USP2a) of USP2.

          Immunogen sequence: YTESARYTD AHYAKSGYGA YTPSSYGANL AASLLEKEKL GFKPVPTSSF LTRPRTYGPS SLLDYDRGRP LLRPDITGGG KRAESQTRGT ERPLGSGLSG GSGFPYGVTN NCLSYLPINA YDQGVTLTQK LDSQSDLARD FSSLRTSDSY RIDPRNLGRS PMLARTRKEL CTLQGLYQTA SCPEYLVDYL ENYGRKGSAS QVPSQAPPSR VPEIISPTYR PIGRYTLWET GK (11-241 aa encoded by BC002955)

          Target Information

          Ubiquitin is a highly conserved protein involved in the regulation of intracellular protein breakdown, cell cycle regulation, and stress response, is released from degraded proteins by disassembly of the polyubiquitin chains. The disassembly process is mediated by ubiquitin-specific proteases (USPs).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 41 kDa ubiquitin-specific protease; Deubiquitinating enzyme 2; Ubiquitin carboxyl-terminal hydrolase 2; ubiquitin specific protease 12; ubiquitin specific protease 2; ubiquitin specific protease 9; Ubiquitin thioesterase 2; ubiquitin thiolesterase 2; Ubiquitin-specific-processing protease 2; Ubiquitin-specific-processing protease testis; UBP-t

          View more View less

          Gene Aliases: B930035K21Rik; MNCb-0190; UBP-t; UBP41; Ubp69; USP2; USP9

          View more View less

          UniProt ID: (Human) O75604, (Mouse) O88623, (Rat) Q5U349

          View more View less

          Entrez Gene ID: (Human) 9099, (Mouse) 53376, (Rat) 115771

          View more View less

          Function(s)
          cysteine-type endopeptidase activity thiol-dependent ubiquitin-specific protease activity protein binding cyclin binding ubiquitin protein ligase binding identical protein binding metal ion binding peptidase activity cysteine-type peptidase activity hydrolase activity thiol-dependent ubiquitinyl hydrolase activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter ubiquitin-dependent protein catabolic process cell cycle muscle organ development protein deubiquitination circadian regulation of gene expression entrainment of circadian clock by photoperiod locomotor rhythm positive regulation of mitotic cell cycle circadian behavior negative regulation of skeletal muscle tissue development positive regulation of skeletal muscle tissue development protein stabilization regulation of transcription, DNA-templated proteolysis proteasome-mediated ubiquitin-dependent protein catabolic process rhythmic process regulation of proteasomal protein catabolic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67f4cd694f-jv9gd:80/100.66.135.11:80.
          git-commit: 961b7666e63bcc533d6299fdd217d6a70bd96cf1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0