Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSQLSSTLKR YTESARYTDA HYAKSGYGAY TPSSYGANLA ASLLEKEKLG FKPVPTSSFL TRPRTYGPSS LLDYDRGRPL LRPDITGGGK RAESQTRGTE RPLGSGLSGG SGFPYGVTNN CLSYLPINAY DQGVTLTQKL DSQSDLARDF SSLRTSDSYR IDPRNLGRSP MLARTRKELC TLQGLYQTAS CPEYLVDYLE NYGRKGSASQ VPSQAPPSRV PEIISPTYRP IGRYTLWETG KGQAPGPSRS SSPGRDGMNS KSAQGLAGLR NLGNTCFMNS ILQCLSNTRE LRDYCLQRLY MRDLHHGSNA HTALVEEFAK LIQTIWTSSP NDVVSPSEFK TQIQRYAPRF VGYNQQDAQE FLRFLLDGLH NEVNRVTLRP KSNPENLDHL PDDEKGRQMW RKYLEREDSR IGDLFVGQLK SSLTCTDCGY CSTVFDPFWD LSLPIAKRGY PEVTLMDCMR LFTKEDVLDG DEKPTCCRCR GRKRCIKKFS IQRFPKILVL HLKRFSESRI RTSKLTTFVN FPLRDLDLRE FASENTNHAV YNLYAVSNHS GTTMGGHYTA YCRSPGTGEW HTFNDSSVTP MSSSQVRTSD AYLLFYELAS PPSRM
Ubiquitin is a highly conserved protein involved in the regulation of intracellular protein breakdown, cell cycle regulation, and stress response, is released from degraded proteins by disassembly of the polyubiquitin chains. The disassembly process is mediated by ubiquitin-specific proteases (USPs).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 41 kDa ubiquitin-specific protease; Deubiquitinating enzyme 2; Ubiquitin carboxyl-terminal hydrolase 2; ubiquitin specific protease 12; ubiquitin specific protease 9; Ubiquitin thioesterase 2; Ubiquitin-specific-processing protease 2
Gene Aliases: UBP41; USP2; USP9
UniProt ID: (Human) O75604
Entrez Gene ID: (Human) 9099
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support