Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: TERKKRRKGL VPGLVNLGNT CFMNSLLQGL SACPAFIRWL EEFTSQYSRD QKEPPSHQYL SLTLLHLLKA LSCQEVTDDE VLDASCLLDV LRMYRWQISS FEEQDAHELF HVITSSLEDE RDRQPRVTHL FDVHSLEQQS EITPKQITCR TRGSPHPTSN HWKSQHPFHG RLTSNMVCKH CEHQSPVRFD TFDSLSLSIP AATWGHPLTL DHCLHHFISS ESVRDVVCDN CTKIEAKGTL NGEKVEHQRT TFVKQLKLGK LPQCLCIHLQ RLSWSSHGTP LKRHEHVQFN EFLMMDIYKY HLLGHKPSQH NPKLNKNPGP TLELQDGPGA PTPV; Positive Samples: Mouse lung, Rat lung; Cellular Location: Mitochondrion outer membrane
USP30, a member of the ubiquitin-specific protease family, is a novel mitochondrial deubiquitinating (DUB) enzyme (Nakamura and Hirose, 2008).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 30; FLJ40511; MGC10702; Ub-specific protease 30; Ubiquitin carboxyl-terminal hydrolase 30; ubiquitin specific protease 30; Ubiquitin thioesterase 30; ubiquitin thiolesterase 30; ubiquitin-specific protease 30; Ubiquitin-specific-processing protease 30
Gene Aliases: 6330590F17Rik; AI851327; D5Ertd483e; USP30
UniProt ID: (Human) Q70CQ3, (Rat) D3ZPG5, (Mouse) Q3UN04
Entrez Gene ID: (Human) 84749, (Rat) 304579, (Mouse) 100756
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support