Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: VCSTSVDGS ECVTLPVYFR ERKSRPSSTS SASALYGQPL LLSVPKHKLT LESLYQAVCD RISRYVKQPL PDEFGSSPLE PGACNGSRNS CEGEDEEEME HQEEGKEQLS ETEGSGEDEP GNDPSETTQK KIKGQPCPKR LFTFSLVNSY GTADINSLAA DGKLLKLNSR STLAMDWDSE TRRLYYDEQE SEAYEKHVSM LQPQKKKKTT VALR (567-779 aa encoded by B C125131)
The protein encoded by this gene is a protease that deubiquitinates target proteins such as ADORA2A and TRIM21. The encoded protein shuttles between the nucleus and cytoplasm and is involved in maintaining operational fidelity in the endoplasmic reticulum. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 4; ubiquitin carboxyl-terminal esterase 4; Ubiquitin carboxyl-terminal hydrolase 4; ubiquitin specific peptidase 4 (proto-oncogene); ubiquitin specific protease 4 (proto-oncogene); Ubiquitin thioesterase 4; ubiquitin thiolesterase 4; ubiquitin-specific processing protease 4; Ubiquitin-specific-processing protease 4; Ubiquitous nuclear protein; Ubiquitous nuclear protein homolog
Gene Aliases: F730026I20Rik; mKIAA4155; UNP; UNPH; USP4
UniProt ID: (Human) Q13107, (Mouse) P35123
Entrez Gene ID: (Human) 7375, (Mouse) 22258
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support