Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: AEEGGVPADE VILVELYPSG FQRSFFDEED LNTIAEGDNV YAFQVPPSPS QGTLSAHPLG LSASPRLAAR EGQRFSLSLH SESKVLILFC NLVGSGQQAS RFGPPFLIR
May recognize and hydrolyze the peptide bond at the C-terminal Gly of ubiquitin. Involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deubiquitinating enzyme 43; Ubiquitin carboxyl-terminal hydrolase 43; ubiquitin specific protease 43; Ubiquitin thioesterase 43; ubiquitin thiolesterase 43; Ubiquitin-specific-processing protease 43
Gene Aliases: USP43
UniProt ID: (Human) Q70EL4
Entrez Gene ID: (Human) 124739
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support