Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MATPPKRRA VEATGEKVLR YETFISDVLQ RDLRKVLDHR DKVYEQLAKY LQLRNVIERL QEAKHSELYM QVDLGCNFFV DTVVPDTSRI YVALGYGFFL ELTLAEALKF IDRKSSLLTE LSNSLTKDSM NIKAHIHMLL EGLRELQGLQ NFPEKPHH (1-157 aa encoded by BC008890)
This gene encodes a novel protein which is highly conserved in mouse. It interacts with the N-terminus of the androgen receptor and plays a role in facilitating receptor-induced transcriptional activation. It is also likely to be involved in tumorigenesis as it is abundantly expressed in tumor tissues. This gene is part of a gene cluster on chromosome Xp11.23. Alternative splicing results in 2 transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Androgen receptor trapped clone 27 protein; ART-27; Protein UXT; RP1-212G6.2; SKP2-associated alpha PFD 1; Ubiquitously expressed transcript protein; ubiquitously-expressed, prefoldin-like chaperone
Gene Aliases: 0910002B17Rik; ART-27; HSPC024; STAP1; UXT
UniProt ID: (Human) Q9UBK9, (Mouse) Q9WTZ0, (Rat) Q63ZY7
Entrez Gene ID: (Human) 8409, (Mouse) 22294, (Rat) 299313
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support