Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • VANGL2 Antibodies

          Proteintech

          VANGL2 Polyclonal Antibody

          13 References
          View all (12) VANGL2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite VANGL2 Polyclonal Antibody

          • Antibody Testing Data (13)
          VANGL2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          VANGL2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          VANGL2 Antibody (21492-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells using 21492-1-AP (VANGL2 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          VANGL2 Antibody in Immunocytochemistry (ICC/IF)
          VANGL2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          VANGL2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          VANGL2 Antibody in Western Blot (WB)
          VANGL2 Antibody in Western Blot (WB)
          VANGL2 Antibody in Western Blot (WB)
          VANGL2 Antibody in Western Blot (WB)
          VANGL2 Antibody in Western Blot (WB)
          VANGL2 Antibody in Western Blot (WB)
          VANGL2 Antibody in Western Blot (WB)
          VANGL2 Antibody in Western Blot (WB)
          VANGL2 Antibody in Immunoprecipitation (IP)
          VANGL2 Antibody in Immunoprecipitation (IP)

          Product Details

          21492-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 7 publications 7 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 4 publications 4 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          VANGL2 Fusion Protein Ag15833 (268-322 aa encoded by BC103920)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.23 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: IQRVAVWIL EKYYHDFPVY NPALLNLPKS VLAKKVSGFK VYSLGEENST NNSTGQ (268-322 aa encoded by BC103920)

          Target Information

          Van Gogh is an alternate name for the Drosophila strabismus protein. Van Gogh like-2 (VANGL2) is a 521 amino acidprotein in vertebrates similar to strabismus 1 of Drosophila and a component of the frizzled-disheveled tissue polarity pathway. It interacts through it's C-terminal region with the N-terminal half of DVL1, DVL2 and DVL3 and is a potent tumor suppressor. It ispredominantly involved in the control of early morphogenesis and patterning of both axial midline structures and the development of neural plate. It plays a crucial role in the regulation of planar cell polarity, particularly in the orientation of stereociliary bundles in the cochlea. VANGL2 is required for polarization and movement of myocardializing cells and seems to act via RHOA signaling to regulate this process. It is expressed in a wide range of tissues.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: loop tail associated protein; Loop-tail protein 1; Loop-tail protein 1 homolog; Loop-tail-associated protein; MGC119403; MGC119404; Strabismus 1; Van Gogh-like protein 2; vang; vang (van gogh)-like 2; vang-like 2 (van gogh, Drosophila); Vang-like protein 2; vangl2 {ECO:0000312|RGD:1309442}

          View more View less

          Gene Aliases: C530001F03Rik; KIAA1215; loop-tail; Lp; LPP1; LTAP; ska17; ska; STB1; STBM; STBM1; strabismus; Vang1l2; VANGL2

          View more View less

          UniProt ID: (Human) Q9ULK5, (Rat) P84889, (Mouse) Q91ZD4

          View more View less

          Entrez Gene ID: (Human) 57216, (Rat) 289229, (Mouse) 93840

          View more View less

          Function(s)
          protein binding molecular_function
          Process(es)
          establishment of planar polarity neural tube closure hair follicle development heart looping membranous septum morphogenesis muscular septum morphogenesis planar cell polarity pathway involved in axis elongation Rho protein signal transduction anterior/posterior pattern specification heparan sulfate proteoglycan biosynthetic process convergent extension involved in neural plate elongation regulation of Wnt signaling pathway glomerulus development regulation of actin cytoskeleton organization somatic stem cell population maintenance nonmotile primary cilium assembly cell migration involved in kidney development post-anal tail morphogenesis dopaminergic neuron axon guidance serotonergic neuron axon guidance wound healing positive regulation of JUN kinase activity apical protein localization establishment or maintenance of epithelial cell apical/basal polarity somatic stem cell division establishment of body hair planar orientation digestive tract morphogenesis convergent extension involved in axis elongation inner ear receptor stereocilium organization orthogonal dichotomous subdivision of terminal units involved in lung branching morphogenesis planar dichotomous subdivision of terminal units involved in lung branching morphogenesis lateral sprouting involved in lung morphogenesis kidney morphogenesis planar cell polarity pathway involved in heart morphogenesis cochlea morphogenesis establishment of planar polarity involved in neural tube closure planar cell polarity pathway involved in neural tube closure biological_process non-canonical Wnt signaling pathway convergent extension involved in organogenesis Wnt signaling pathway, planar cell polarity pathway inner ear receptor cell development cochlea development regulation of establishment of planar polarity patterning of blood vessels outflow tract septum morphogenesis epicardial cell to mesenchymal cell transition multicellular organism development cardiac vascular smooth muscle cell differentiation establishment of epithelial cell polarity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67f4cd694f-jv9gd:80/100.66.135.11:80.
          git-commit: 961b7666e63bcc533d6299fdd217d6a70bd96cf1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0