Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • AOC3 Antibodies
          The PeproTech website has now moved to Thermofisher.com. All orders for PeproTech products must be placed on thermofisher.com or offline via phone, fax, email, or distributor partner. For any questions, please contact PeproTech.CustomerService@thermofisher.com

          Invitrogen

          VAP-1 Polyclonal Antibody, Biotin, PeproTech®

          View all (22) AOC3 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite VAP-1 Polyclonal Antibody, Biotin, PeproTech®

          • Antibody Testing Data (3)
          VAP-1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          VAP-1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          VAP-1 Antibody (500-P326BT-25UG) in WB

          Western Blot: To detect Human VAP-1 by Western Blot analysis, VAP-1 Polyclonal Antibody, Biotin (Product # 500-P326BT-1MG) can be used at a concentration of 0.1-0.2 µg/mL. When used in conjunction with compatible development reagents, the detection limit for Recombinant Human VAP-1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          VAP-1 Antibody in Western Blot (WB)
          VAP-1 Antibody in Western Blot (WB)
          VAP-1 Antibody in ELISA (ELISA)

          Product Details

          500-P326BT-25UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.2 µg/mL
          -

          ELISA (ELISA)

          0.25-1.0 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived Recombinant Human VAP-1
          View immunogen

          Conjugate

          Biotin Biotin Biotin

          Form

          Lyophilized

          Concentration

          0.1-1.0 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2930432

          Product Specific Information

          AA Sequence of recombinant protein: GRGGDGGEPS QLPHCPSVSP SAQPWTHPGQ SQLFADLSRE ELTAVMRFLT QRLGPGLVDA AQARPSDNCV FSVELQLPPK AAALAHLDRG SPPPAREALA IVFFGRQPQP NVSELVVGPL PHPSYMRDVT VERHGGPLPY HRRPVLFQEY LDIDQMIFNR ELPQASGLLH HCCFYKHRGR NLVTMTTAPR GLQSGDRATW FGLYYNISGA GFFLHHVGLE LLVNHKALDP ARWTIQKVFY QGRYYDSLAQ LEAQFEAGLV NVVLIPDNGT GGSWSLKSPV PPGPAPPLQF YPQGPRFSVQ GSRVASSLWT FSFGLGAFSG PRIFDVRFQG ERLVYEISLQ EALAIYGGNS PAAMTTRYVD GGFGMGKYTT PLTRGVDCPY LATYVDWHFL LESQAPKTIR DAFCVFEQNQ GLPLRRHHSD LYSHYFGGLA ETVLVVRSMS TLLNYDYVWD TVFHPSGAIE IRFYATGYIS SAFLFGATGK YGNQVSEHTL GTVHTHSAHF KVDLDVAGLE NWVWAEDMVF VPMAVPWSPE HQLQRLQVTR KLLEMEEQAA FLVGSATPRY LYLASNHSNK WGHPRGYRIQ MLSFAGEPLP QNSSMARGFS WERYQLAVTQ RKEEEPSSSS VFNQNDPWAP TVDFSDFINN ETIAGKDLVA WVTAGFLHIP HAEDIPNTVT VGNGVGFFLR PYNFFDEDPS FYSADSIYFR GDQDAGACEV NPLACLPQAA ACAPDLPAFS HGGFSHN.

          Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human VAP-1. Anti-Human VAP-1-specific antibody was purified by affinity chromatography and then biotinylated.

          Sandwich ELISA: To detect Human VAP-1 by sandwich ELISA (using 100 µL/well), a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human VAP-1 (500-P326) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human VAP-1.

          Western Blot: To detect Human VAP-1 by Western Blot analysis, this antibody can be used at a concentration of 0.1-0.2 µg/mL. When used in conjunction with compatible development reagents, the detection limit for Recombinant Human VAP-1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

          500-P326BT-1MG will be provided as 2 x 500 µg

          Target Information

          Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9-kb downstream.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Amine oxidase [copper-containing] 3; Amine oxidase copper-containing 3; amine oxidase, copper containing 3 (vascular adhesion protein 1); Copper amine oxidase; HPAO; placenta copper monamine oxidase; Semicarbazide-sensitive amine oxidase; SSAO; VAP-1; Vascular adhesion protein 1

          View more View less

          Gene Aliases: AOC3; HPAO; SSAO; VAP-1; VAP1

          View more View less

          UniProt ID: (Human) Q16853

          View more View less

          Entrez Gene ID: (Human) 8639

          View more View less

          Function(s)
          copper ion binding calcium ion binding protein binding primary amine oxidase activity protein homodimerization activity protein heterodimerization activity quinone binding tryptamine:oxygen oxidoreductase (deaminating) activity aminoacetone:oxygen oxidoreductase(deaminating) activity aliphatic-amine oxidase activity phenethylamine:oxygen oxidoreductase (deaminating) activity
          Process(es)
          xenobiotic metabolic process inflammatory response cell adhesion amine metabolic process response to antibiotic oxidation-reduction process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-f7m9l:80/100.66.129.252:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0