Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Vinculin Antibodies

          Abnova

          VCL Monoclonal Antibody (3F8-1D4)

          View all (55) Vinculin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite VCL Monoclonal Antibody (3F8-1D4)

          • Antibody Testing Data (1)
          VCL Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          VCL Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          VCL Antibody (H00007414-M02) in WB

          VCL monoclonal antibody (M02), clone 3F8-1D4. Western Blot analysis of VCL expression in HeLa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          VCL Antibody in Western Blot (WB)

          Product Details

          H00007414-M02

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -

          ELISA (ELISA)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2b, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3F8-1D4

          Immunogen

          VCL (AAH39174.1, 1 a.a. approximately 1066 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          Immunogen sequence: MPVFHTRTIE SILEPVAQQI SHLVIMHEEG EVDGKAIPDL TAPVAAVQAA VSNLVRVGKE TVQTTEDQIL KRDMPPAFIK VENACTKLVQ AAQMLQSDPY SVPARDYLID GSRGILSGTS DLLLTFDEAE VRKIIRVCKG ILEYLTVAEV VETMEDLVTY TKNLGPGMTK MAKMIDERQQ ELTHQEHRVM LVNSMNTVKE LLPVLISAMK IFVTTKNSKN QGIEEALKNR NFTLEKMSAE INEIIRVLQL TSWDEDAWAS KDTEAMKRAL ASIDSKLNQA KGWLRDPSAS PGDAGEQAIR QILDEAGKVG ELCAGKERRE ILGTCKMLGQ MTDQVADLRA RGQGSSPVAM QKAQQVSQGL DVLTAKVENA ARKLEAMTNS KQSIAKKIDA AQNWLADPNG GPEGEEQIRG ALAEARKIAE LCDDPKERDD ILRSLGEISA LTSKLADLRR QGKGDSPEAR ALAKQVATAL QNLQTKTNRA VANSRPAKAA VHLEGKIEQA QRWIDNPTVD DRGVGQAAIR GLVAEGHRLA NVMMGPYRQD LLAKCDRVDQ LTAQLADLAA RGEGESPQAR ALASQLQDSL KDLKARMQEA MTQEVSDVFS DTTTPIKLLA VAATAPPDAP NREEVFDERA ANFENHSGKL GATAEKAAAV GTANKSTVEG IQASVKTARE LTPQVVSAAR ILLRNPGNQA AYEHFETMKN QWIDNVEKMT GLVDEAIDTK SLLDASEEAI KKDLDKCKVA MANIQPQMLV AGATSIARRA NRILLVAKRE VENSEDPKFR EAVKAASDEL SKTISPMVMD AKAVAGNISD PGLQKSFLDS GYRILGAVAK VREAFQPQEP DFPPPPPDLE QLRLTDELAP PKPPLPEGEV PPPRPPPPEE KDEEFPEQKA GEVINQPMMM AARQLHDEAR KWSSKGNDII AAAKRMALLM AEMSRLVRGG SGTKRALIQC AKDIAKASDE VTRLAKEVAK QCTDKRIRTN LLQVCERIPT ISTQLKILST VKATMLGRTN ISDEESEQAT EMLVHNAQNL MQSVKETVRE AEAASIKIRT DAGFTLRWVR KTPWYQ

          Target Information

          Vinculin (VCL) is a cytoskeletal protein associated with cell-cell and cell-extracellular matrix adherens-type junctions. It functions as one of several interacting proteins involved in anchoring F-actin to the membrane. It has been shown that a sequence of molecular interactions might be involved in the transmembrane assembly of adhesion plaques. In the assembly of adhesion plaques, the beta subunit of integrin binds to talin. Talin binds to vinculin that interacts with alpha-actinin and possibly with itself. Since alpha-actinin binds to and cross-links actin filaments, vinculin represents a key element in the transmembrane linkage of the extracellular matrix to the cytoplasmic microfilament system. Mutations in the gene can result in cardiomyopathy dilated 1W or familial hypertrophic 15.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: epididymis luminal protein 114; Metavinculin; MV; RP11-178G16.3; vinc; Vinculin

          View more View less

          Gene Aliases: CMD1W; CMH15; HEL114; MV; MVCL; VCL

          View more View less

          UniProt ID: (Human) P18206

          View more View less

          Entrez Gene ID: (Human) 7414

          View more View less

          Function(s)
          dystroglycan binding actin binding structural molecule activity protein binding beta-catenin binding ubiquitin protein ligase binding alpha-catenin binding cadherin binding cadherin binding involved in cell-cell adhesion
          Process(es)
          morphogenesis of an epithelium platelet degranulation movement of cell or subcellular component muscle contraction cell adhesion cell-matrix adhesion lamellipodium assembly negative regulation of cell migration adherens junction assembly protein localization to cell surface apical junction assembly axon extension platelet aggregation epithelial cell-cell adhesion
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0