Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MERPDKAAL NALQPPEFRN ESSLASTLKT LLFFTALMIT VPIGLYFTTK SYIFEGALGM SNRDSYFYAA IVAVVAVHVV LALFVYVAWN EGSRQWREGK QD (1-101 aa encoded by BC105694)
Required for the assembly of the V0 complex of the vacuolar ATPase (V-ATPase) in the endoplasmic reticulum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Myopathy with excessive autophagy protein; Vacuolar ATPase assembly integral membrane protein VMA21; VMA21 vacuolar H+-ATPase homolog (S. cerevisiae)
Gene Aliases: 2610030H06Rik; AI840175; MEAX; RGD1566155; VMA21; XMEA
UniProt ID: (Human) Q3ZAQ7, (Mouse) Q78T54
Entrez Gene ID: (Human) 203547, (Rat) 501658, (Mouse) 67048
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support